missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Lead Tin ingot (Rose's metal)

Product Code. 11388498
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11388498

Brand: Thermo Scientific Alfa Aesar 040516.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56001-37-7 | C10H14N5Na2O14P3 | 567.14 g/mol

Rose's metal was used as the droplet fluid, because it has a low melting point (e460 K). Rose's metal is used as a sample in computer-controlled diffractometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56001-37-7
Molecular Formula C10H14N5Na2O14P3
Molecular Weight (g/mol) 567.14
MDL Number MFCD03410297
InChI Key FIZIYLKEXVIRHJ-KHRSEZDTNA-L
Synonym guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
PubChem CID 131676145
IUPAC Name ({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point 95°C to 96°C
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn; 50:25:25 wt%
Quantity 500 g
Form Melting Alloy
Solubility Information Insoluble in water.
Assay (Rose's metal)
Chemical Name or Material Bismuth Lead Tin ingot (Rose's metal)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.