missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UN Number | UN3089 |
Quantity | 100 g |
Percent Purity | 99.5% |
Mesh Size | −325 Mesh |
Odor | Odorless |
Weight | Apparent density: 3.2g/cm3 to 4.0g/cm3 |
Form | Powder |
Assay | (metals basis) |
Chemical Name or Material | Bismuth powder |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Bismuth powder
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'