missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Standard For ICP, 1000μg/mL in 10% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog Number 11408649
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7440-69-9, 7697-37-2, 7732-18-5
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
CHEBI:33301
bismuth
[Bi]
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7440-69-9, 7697-37-2, 7732-18-5
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
CHEBI:33301
bismuth
[Bi]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Bi1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 10.0, 89.9
Quantity 125 mL
CAS Max % 0.1, 10.0, 89.9
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
10% HNO3 1000μg/mL PLBI4-2Y, -2X
Solvent or Matrix 10% HNO3
Chemical Name or Material Bismuth Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'