missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subnitrate EU Pharmacopoeia, Fisher Chemical™

Product Code. 12990364
Click to view available options
Quantity:
500 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Concentration 10μg/mL
CAS 28434-01-7
Packaging Ampoule
Quantity 500 mL
Molecular Formula C22H26O3
InChI Key VEMKTZHHVJILDY-UXHICEINSA-N
SMILES CC(=CC1C(C1(C)C)C(=O)OCC2=COC(=C2)CC3=CC=CC=C3)C
IUPAC Name (5-benzylfuran-3-yl)methyl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
Molecular Weight (g/mol) 338.447
PubChem CID 162381
ChEBI CHEBI:39280
Grade EU Ph Ref: 1011502
Chemical Name or Material Bismuth subnitrate solution.
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth subnitrate solution. European Pharmacopoeia Ref: 1011502
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands, forearms andface thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'