missing translation for 'onlineSavingsMsg'
Learn More

Bismuth subsalicylate, 96%

Product Code. 11383978
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11383978

Brand: Thermo Scientific Alfa Aesar 040395.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14882-18-9 | C7H6BiO4 | 363.101 g/mol

To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm^3Bismuth subsalicylate is used to treat temporary discomforts of the stomach and gastrointestinal tract, such as diarrhea, indigestion, heartburn and nausea. It also acts as an antacid. It is practiced to decrease the number of bowel movements and to make the stool firmer. It is also used to reduce inflammation within the intestine. It is further used in growth media for selective isolation of microorganisms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14882-18-9
Molecular Formula C7H6BiO4
Molecular Weight (g/mol) 363.101
MDL Number MFCD00085368
InChI Key QBWLKDFBINPHFT-UHFFFAOYSA-L
Synonym bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum
PubChem CID 16682734
IUPAC Name 1,3,2$l^{2}-benzodioxabismin-4-one;hydrate
SMILES C1=CC=C2C(=C1)C(=O)O[Bi]O2.O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 96%
Solubility Information Practically insoluble in water,alcoholSoluble in acid and alkali. Insoluble in cold water,ethanol,diethyl ether and n-octanol.
Formula Weight 362.09
Odor Odorless
Appearance White powder
Chemical Name or Material Bismuth subsalicylate
compliance-icons
Product Identifier
  • Bismuth subsalicylate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.