missing translation for 'onlineSavingsMsg'
Learn More

Bismuth titanium isopropoxide, 5% w/v in isopropanol, Thermo Scientific™

Product Code. 11390048
25 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Preparation of thin film mixed oxides of Ti and Bi

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338391-61-0
Molecular Formula C21H49BiO7Ti
MDL Number MFCD01865878
Synonym bismuthane titanium, bismuth-titanium 1/1, bismuth, compd. with titanium 1:1, bismuth titanium isopropoxide w/v in isopropanol, bismuth titanium isopropoxide w/v in isopropanol, packaged under argon in resealable chemseal bottles

Specifications

Color Yellow
Linear Formula BiTi[OCH(CH3)2]7
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 670.48
Physical Form Liquid
Chemical Name or Material Bismuth titanium isopropoxide
compliance-icons compliance-icons
Product Identifier
  • Bismuth titanium isopropoxide, 5% w/v in isopropanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Bismuth titanium isopropoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth titanium isopropoxide, 5% w/v in isopropanol, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.