missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisphenol A diglycidyl ether resin, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.
Solubility
Soluble in 100% ethanol, dimethyl sulfoxide (100 mM), dimethyl formamide, chloroform, methanol, and ethanol (50 mM). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.
Solubility
Soluble in 100% ethanol, dimethyl sulfoxide (100 mM), dimethyl formamide, chloroform, methanol, and ethanol (50 mM). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.17 |
Refractive Index | 1.573 |
Quantity | 25 g |
Beilstein | 299026 |
Solubility Information | Soluble in 100% ethanol,dimethyl sulfoxide (100 mM),dimethyl formamide,chloroform,methanol,and ethanol (50 mM). Insoluble in water. |
Formula Weight | 340.42 |
Chemical Name or Material | Bisphenol A diglycidyl ether |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'