Learn More
Moldex™ BITREX™ Fit Test Solution
For use with BITREX Fit Test Kit
Marque: Moldex™ 50401
Code : NEW
Informations supplémentaires : Weight : 0.04512kg
Description
- Comply with OSHA standards for qualitative fit testing
- Respirators sold separately
Spécification
Fit Test Solution | |
BITREX Fit Test Kit | |
6 x 2.5 mL Ampules |