missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Black 7940 Series Thermal Transfer Printer Ribbon
Ribbon for Brady Printer BBP11 and BBP12
Brand: Brady™ 804459
26.71 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- General purpose, standard performance wax ribbon for paper and temporary labelling
Specifications
Black | |
65 mm | |
Black 7940 Series Thermal Transfer Printer Ribbon | |
1 Ea. |
70 m | |
12.7 mm I.D. | |
Ink side out | |
R7940-65X70/O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction