missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Black 96-Well Immuno Plates
BIOPHARMA

Product Code. p-4555272
Click to view available options
Surface Treatment:
High Binding
MaxiSorp
Medium Binding
PolySorp
Universal Binding
Volume (Metric) Working:
280 μL
350 μL
Unit Size:
Pack of 50
Pack of 80

Product Code. 10030581

Brand: Thermo Scientific™ 437111

Only null left
Add to basket
Edge
Add to basket

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.

Features:

  • Plate has SBS footprint — fits standard equipment
  • Flat-bottom wells
  • Certified binding homogeneity/reproducibility

Choice of surface:

  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
  • Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.

Compatible with:

Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering Information:

Availability may vary by country

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Hydrophilic
Format Standard
Lid Without Lid
Detection Method Fluorescence
Array 8 x 12
Packaging 80 Cases
Binding Type Adsorption
Color Black
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 350 μL
Surface Treatment MaxiSorp
Well Shape Flat Bottom (F-well)
For Use With (Application) Fluorescence
No. per Case 80
No. per Pack 10
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design F96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.