missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Black Eriochrome T, Chem Lab™
Specifications
Solubility Information | Soluble in water |
---|---|
Odor | Specific |
Color | Black |
Quantity | 1 L |
Physical Form | Liquid |
Specifications
Soluble in water | |
Black | |
Liquid |
Specific | |
1 L |