Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Brand: Axygen™ HT-300-CBK-HTR-S-5
This item is not returnable.
View return policy
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- Available in non-sterile, sterile, nonfiltered, and filtered
- Available as Clear or Conductive black polypropylene tips
- Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
300 μL | |
Sterile | |
Not Autoclavable | |
Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
Automation | |
Hamilton MICROLAB Nimbus |
Non-filtered | |
Black | |
DNase, RNase and Pyrogen free | |
Hanging Tip Rack | |
5 Racks/Pk, 12 Pks/Cs. | |
Polypropylene |