Learn More
Fisherbrand™ Black Phenolic PolyCone Cap
Product Code. 15573261
Click to view available options
Quantity:
1600/Cs.
2200/Cs.
4100/Cs.
5500/Cs.
576/Cs.
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
Specifications
Color Black
Material Phenolic Resin
Quantity 576/Cs.
Closure Color Black
Liner Material PolyCone
Neck Finish 38-400
Closure Included Yes
Product Type PolyCone Lined Cap
Closure Liner Polyseal
Closure Material Phenolic
Closure Size 38-400 mm
Closure Type Screw Cap