missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Black Plastic Bottle Screw Caps

Product Code. 11568212
Click to view available options
Closure Size:
20-400 mm
28-400 mm
33-400 mm
Liner Material:
Rubber
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color Black
Description Screw cap
Material Plastic
Length (Metric) 80mm
No. per Case 576 Cs.
Quantity 576/PK.
Sterility Sterile
Liner Material Rubber
Neck Finish 28-400
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) Bottle
Closure Liner 14B Rubber
Closure Material Plastic
Closure Size 28-400 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'