missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly

Provides for accurate, reliable dispensing of various samples. Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly are manufactured from clear Type 1 neutral glass.

£42.18 - £44.12

Specifications

Color Clear
Diameter (Metric) Outer 20 mm
Material Borosilicate Glass
Neck Style Screw
Thread Style Screw
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
14883572
View Documents
Fisherbrand™
800059
10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
14863572
missing translation for 'viewDocuments'
Fisherbrand™
800058
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • With screw neck
  • With black caps and bulbs
  • Droppers are assembled but not fitted to vials
  • Dropper assembly comprised of clear glass bottle dropper tube fitted to a polypropylene cap with a natural rubber bulb
Specifications

Specifications

Clear
Borosilicate Glass
Screw
Dropper Screw Cap
Screw Thread Vial
20 mm
Screw
Polypropylene
40/Pk.
Videos
SDS
Documents

Documents

Certificates