Learn More
Grant Instruments™ Blanket for Inspissator
Provides thermal insulation and minimizes drafts. Grant Instruments™ Blanket for Inspissator is insect resistant for use with production of tuberculosis culture medium
Brand:  Grant Instruments™ BIN
Code : 50
Additional Details : Weight : 2.00000kg
Product Code. 11410998
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For use with T100IN Inspissator
  • Insect resistant
  • Provides thermal insulation and minimizes drafts when placed over the containers
Specifications
Specifications
Inspissator