missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blasticidin S Hydrochloride (From Streptomyces Griseochromogenes), Fisher BioReagents
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass
Specifications
Specifications
Type | Blasticidin S Hydrochloride |
Physical Form | Powder/Solid |
Melting Point | 248°C |
Assay Percent Range | ≥99.0 % |
Packaging | Amber Glass |
Potency | 900μg/mg min. |
Formula Weight | 458.69 |
Percent Purity | 99.0% min. |
Color | White |
Quantity | 100 mg |
Show More |
Safety and Handling
Product Identifier
- Blasticidin S Hydrochloride
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H300-Fatal if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'