missing translation for 'onlineSavingsMsg'
Learn More

Blasticidin S Hydrochloride (From Streptomyces Griseochromogenes), Fisher BioReagents

Product Code. 10658203
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass

Chemical Identifiers

CAS 3513-03-9
Molecular Formula C17H27ClN8O5
Molecular Weight (g/mol) 458.90
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
Synonym BLA-S, Cytovirin
PubChem CID 124080960
IUPAC Name (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
SMILES CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl

Specifications

Type Blasticidin S Hydrochloride
Physical Form Powder/Solid
Melting Point 248°C
Assay Percent Range ≥99.0 %
Packaging Amber Glass
Potency 900μg/mg min.
Formula Weight 458.69
Percent Purity 99.0% min.
Color White
Quantity 50 mg
Chemical Name or Material Blasticidin S Hydrochloride, From Streptomyces griseochromogenes
Show More Show Less
compliance-icons
Product Identifier
  • Blasticidin S Hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'