Learn More
BRAND™ Blaubrand™ Borosilicate Glass Class A USP Volumetric Flask
Manufactured from Class A Borosilicate Glass and complies with DIN EN ISO 1042 standards.BRAND™ Blaubrand™ Borosilicate Glass Class A USP Volumetric Flask includes polypropylene stopper and USP batch certificate. DE-M Marking provides easy volume check.
£33.00 - £67.40
Specifications
Calibration | Calibrated To Contain |
---|---|
Certifications/Compliance | DIN EN ISO 1042, USP batch certificate, USP individual certificate upon request |
Closure Material | Polypropylene |
Color | Clear |
Material | Borosilicate Glass |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
11814180
|
BRAND™
36938 |
5 mL | 10/19 |
£33.00
Pack of 2 |
In Stock
Log in to see stock. |
||||
10113352
|
BRAND™
36948 |
50 mL | 12/21 |
£36.45
Pack of 2 |
In Stock
Log in to see stock. |
||||
15527841
|
BRAND™
36945 |
20 mL | 10/19 |
£33.00
Pack of 2 |
In Stock
Log in to see stock. |
||||
11824180
|
BRAND™
36943 |
10 mL | 10/19 |
£39.60
Pack of 2 |
In Stock
Log in to see stock. |
||||
10163542
|
BRAND™
36947 |
25 mL | 10/19 |
£38.70
Pack of 2 |
In Stock
Log in to see stock. |
||||
10778904
|
BRAND™
36949 |
100 mL | 14/23 |
£64.90
Pack of 2 |
In Stock
Log in to see stock. |
||||
10663543
|
BRAND™
36950 |
200 mL | 14/23 |
£45.25
Pack of 2 |
In Stock
Log in to see stock. |
||||
10081382
|
BRAND™
36951 |
250 mL | 14/23 |
£48.50
Pack of 2 |
In Stock
Log in to see stock. |
||||
10346522
|
BRAND™
36952 |
500 mL | 19/26 |
£57.64
Pack of 2 |
In Stock
Log in to see stock. |
||||
10356522
|
BRAND™
36953 |
1000 mL | 24/29 |
£49.25
Each |
In Stock
Log in to see stock. |
||||
10521623
|
BRAND™
36954 |
2000 mL | 29/32 |
£67.40
Each |
In Stock
Log in to see stock. |
||||
Description
- Manufactured from Class A Borosilicate Glass.
- Complies with DIN EN ISO 1042 standards.
- Polypropylene stopper included.
- DE-M Marking provides easy volume check.
- Calibrated to contain. (TC, In)
- USP batch certificate included. USP individual certificate available on request.
Specifications
Calibrated To Contain | |
Polypropylene | |
Borosilicate Glass |
DIN EN ISO 1042, USP batch certificate, USP individual certificate upon request | |
Clear | |
A |