Learn More
Brand™ Blaubrand™ Amber Borosilicate Glass Class A USP Volumetric Flask With Glass Stopper
Manufactured from Amber Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Amber Borosilicate Glass Class A USP Volumetric Flask with Glass Stopper protects light-sensitive liquids from deterioration. Glass stopper is Interchangeable with polypropylene stopper. USP individual certificate available upon request.
Brand:  Brand™ 37464
 View more versions of this product
Product Code. 15252845
  • £79.75 / Pack of 2
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Amber Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Includes amber glass stopper, interchangeable with polypropylene stopper.
  • Dark amber flask protects light-sensitive liquids from deterioration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibrated To Contain
Glass
25 mL
±0.03 mL
Volumetric Flask
A
DIN EN ISO 1042, USP individual certificate by request
Amber
Borosilicate Glass
2 Piece
±0.03 mL
10/16