missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BLAUBRAND™ Automatic Burette with Bottle and Intermediate Stopcock

Pellet pattern automatic burette includes 2000mL soda lime glass bottle and Schellbach lateral needle-valve stopcock PTFE-key. Graduations are within the error limits defined by DIN and ISO and carry the DE-M marking.

Brand:  BRAND™ 22768

 View more versions of this product

Product Code. 10180561

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Borosilicate glass construction (Boro 3.3) to DIN EN ISO 385.
  • Calibrated to deliver (TD, Ex).
  • Schellbach stripe.
  • Includes needle-valve stopcock with PTFE spindle (intermediate stopcock with PTFE-key).
  • Automatic zeroing.
  • Intermediate stopcock 4 NS/19 for recirculating residual liquid.
  • Total height approx. 1m incl. 2000mL bottle (soda-lime glass).
  • All BLAUBRAND™ automatic burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL
Borosilicate Glass, PTFE
±0.05 mL
DIN EN ISO 385
Bottle, Needle-valve stopcock, PTFE spindle
0.1 mL
Automatic Buret
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers