Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Product Code. p-8000978 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
Unit Size:
Each
Product Code. 10663561
Brand: BRAND™ 43308
Only null left
Add to basket
Add to basket
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
  • Calibrated density bottle made with Borosilicate glass
  • DIN ISO 3507, type Gay-Lussac
  • Calibrated to contain (TC, In)
  • Includes individual certificate
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • The volume in cm³ is specified to a precision of 3 decimal places
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2Pack
Capacity (Metric) 10 mL
Material Borosilicate Glass
Product Type Calibrated Density Bottle