Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Compact Burettes
Fast to dismantle and easy to clean. BRAND™ Blaubrand™ Borosilicate Glass Compact Burettes are also simple to repair because all individual components replaceable.
Brand:  BRAND™ 13918
 View more versions of this product
Product Code. 10464342
  • £85.20 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes removable PTFE stopcock
  • Fast to dismantle and easy to clean.
  • Simple repair - all individual components replaceable.
  • Borosilicate glass construction (Boro 3.3 to DIN EN ISO 385.)
  • Calibrated to deliver (TD, Ex).
  • Schellbach stripe.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
Borosilicate Glass
Removable PTFE Stopcock
±0.05 mL
DIN EN ISO 385, USP
800 mm
0.1 mL
PTFE
Class AS