missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BlauBRAND™ Burette Tube for Compact Burette
Borosilicate glass construction. Replacement burette tube for compact burette.
Brand: BRAND™ 13948
29.75 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- BLAUBRAND™ borosilicate glass (3.3) construction
- Replacement burette tube
Specifications
50 mL | |
Compact Burette | |
Borosilicate Glass | |
Buret |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction