missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper is available with narrow neck or wide neck in a variety of sizes. DE-M marking on the neck provides quick and accurate volume check. Polypropylene stopper included.
£30.50 - £282.77
Specifications
Calibration | Calibrated To Contain |
---|---|
Certifications/Compliance | DIN EN ISO 1042 |
Closure Material | Polypropylene |
Color | Clear |
Description | DE-M Marking |
Product Code | Brand | Capacity (Metric) | Neck Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Neck Style | Price | Quantity & Availability | ||||
11854150
|
BRAND™
37249 |
100 mL | Narrow |
£38.44
Pack of 2 |
In Stock
Log in to see stock. |
||||
11844150
|
BRAND™
37291 |
100 mL | Wide |
£61.87
Pack of 2 |
In Stock
Log in to see stock. |
||||
11693199
|
BRAND™
37243 |
10 mL | Wide |
£30.50
Pack of 2 |
In Stock
Log in to see stock. |
||||
11884010
|
BRAND™
37250 |
200 mL | Narrow |
£98.40
Pack of 2 |
In Stock
Log in to see stock. |
||||
11333884
|
BRAND™
37290 |
50 mL | Wide |
£35.81
Pack of 2 |
In Stock
Log in to see stock. |
||||
11814150
|
BRAND™
37251 |
250 mL | Narrow |
£45.87
Pack of 2 |
In Stock
Log in to see stock. |
||||
10120571
|
BRAND™
37255 |
5 L | Narrow |
£282.77
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
11858361
|
BRAND™
37252 |
500 mL | Narrow |
£70.12
Pack of 2 |
In Stock
Log in to see stock. |
||||
11864010
|
BRAND™
37247 |
25 mL | Narrow |
£32.15
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10211601
|
BRAND™
37253 |
1000 mL | Narrow |
£60.00
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
10488492
|
BRAND™
37254 |
2000 mL | Narrow |
£84.57
Each |
In Stock
Log in to see stock. |
||||
11673199
|
BRAND™
37234 |
1000 mL | Wide |
£50.95
Each |
In Stock
Log in to see stock. |
||||
11683199
|
BRAND™
37238 |
5 mL | Wide |
£35.30
Pack of 2 |
In Stock
Log in to see stock. |
||||
10287863
|
BRAND™
37245 |
20 mL | Wide |
£74.25
Pack of 2 |
In Stock
Log in to see stock. |
||||
11854010
|
BRAND™
37246 |
20 mL | Narrow |
£31.15
Pack of 2 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
11874010
|
BRAND™
37248 |
50 mL | Narrow |
£40.10
Pack of 2 |
In Stock
Log in to see stock. |
||||
11844020
|
BRAND™
37293 |
25 mL | Wide |
£58.80
Pack of 2 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- DAkkS calibration certificate available upon request
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- DE-M marking provides quick and accurate volume check
- Polypropylene stopper included
Specifications
Calibrated To Contain | |
Polypropylene | |
DE-M Marking | |
Yes | |
Stopper |
DIN EN ISO 1042 | |
Clear | |
Borosilicate Glass | |
A |