Learn More
Brand™ Blaubrand™ Eterna™ Class AS Bulb Pipets, with Certificate
Manufactured from AR-GLAS™ Soda-lime glass and complies with DIN EN ISO 648 standards. Delivered with individual conformity certificate.
Brand:  Brand™ 930512
34.97 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 15815532
  • £40.90 / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from AR-GLAS™ Soda-lime glass.
  • DIN EN ISO 648. Calibrated to deliver (TD, Ex).
  • Waiting time 5 seconds.
  • Delivered with batch number and one batch certificate per packing unit of the manufacturer.
  • Delivered with individual conformity certificate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
To Deliver
AS
Yes
DE-M Marking
Red
Volumetric Bulb Pipet
DIN EN ISO 648, DIN 12691
AR-GLAS, Soda Lime Glass
Amber
0.02 mL
10 mL