missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, with Certificate

High contrast blue enamel represents optimum combination of resistance and readability. Delivered with individual USP conformity certificate.

Brand:  BRAND™ 950601

36.27 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15865562

  • £44.95 / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of AR-GLAS™
  • Optimized flow geometry in tip ensures reliable liquid dispensing from pipet
  • Fire-polished edges for more durability
  • Available in all ISO sizes from 0.5 to 100mL
  • Delivered with individual USP conformity certificate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP
AR-GLAS
Bulb Pipet
A
0.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers