Learn More
Syngene Blended Carbon Filter
Blended carbon filter for inorganics and organics.
Brand: Syngene SYN-ACF411H
Additional Details : Weight : 12.70050kg
Description
- Spare Part
Specifications
Carbon | |
4 in. heavy duty blended layered carbon filter | |
Activated Carbon | |
Chemical extraction |
Airclean | |
Carbon Filter | |
ACF411H | |
Silver |