missing translation for 'onlineSavingsMsg'
Learn More

SCAT Blind Plugs for HPLC

Product Code. 13233859 Shop All SCAT Europe Products
Click to view available options
For Use With:
Exhaust Filter
Ground Neck Bottles w/ Locknut
Safety Caps, Safety Waste Caps
Color:
Colorless
White
Material:
PFA
PTFE
Thread Size:
GL14
NPT 1/8 in.
NS29/32
UNF 1/4 in. 28G
Unit Size:
Each
Pack of 10

Product Code. 13233859

Brand: SCAT 160501

Only null left
Add to basket
Add to basket

Blind plugs for use with HPLC instruments.

Be prepared for HPLC with these SCAT™ blind plugs.

Specifications

Type Blind Plug
Color Colorless
Diameter (Metric) Outer 6.35 mm
Autoclavable Autoclavable
For Use With (Application) HPLC/UHPLC Systems
Packaging Quantity 10 Pk.
Thread Size UNF 1/4 in. 28G
For Use With Safety Caps, Safety Waste Caps
Material PFA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.