Learn More
OHAUS™ Block Cover
OHAUS™ Block Cover is a clear cover for use with low temperature blocks.
£115.00 - £120.00
Specifications
Type | Accessory, Block Cover |
---|---|
For Use With (Application) | Low Temperatures |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15712599
|
OHAUS™
30400179 |
Cover Low-Temperature 2 Block |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
15722599
|
OHAUS™
30400180 |
Cover Low-Temperature 4 Block |
£115.00
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15732599
|
OHAUS™
30400181 |
Cover Low-Temperature 6 Block |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- Block heater accessories include modular blocks to accommodate almost any sample
- Modular blocks feature close contact of tubes-to-walls allowing for maximum heat retention
- Most blocks feature a thermometer well for measuring block temperature
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Accessory, Block Cover | |
Low Temperatures |