missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control
BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control
Brand: Invitrogen™ 14750100
Code : 50
Additional Details : Weight : 0.01050kg
Description
The BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control is an ideal indicator of lipid-mediated transfection efficiency for RNAi experiments, particularly those using the Lipofectamine™ RNAiMAX Transfection Reagent. The sequence of the BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control is not homologous to any known gene, preventing induction of nonspecific cellular events caused by introduction of the duplex into cells. The BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control provides:Specifications
The BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control is provided at a concentration of 20 μM. Store at -20°C. Guaranteed stable for 12 months when properly stored. |
|
Fluorescent | |
RNA Transfection | |
Alexa Fluor™, BLOCK-iT™ | |
2 x 125 μL | |
Dry Ice |
Transfection Control | |
RNA | |
Alexa Fluor 555 | |
Red fluorescent transfection control | |
siRNA |
For Research Use Only. Not for use in diagnostic procedures.