Learn More
Thermo Scientific™ Blocker™ BSA
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Brand:  Thermo Scientific™ 37520
136.32 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 11889310
  • £192.00 / 125mL
Explore more special offers
Description
Description
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blocker BSA in TBS (10X)
125 mL
Western Blot, ELISA, Immunohistochemistry (IHC)
10% BSA
Single purified protein
Blocker BSA
Liquid
Blocker™
TBS
Store at 4°C.