missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Product Code. 10496134 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Chemical Name or Material:
PBS
TBS
Unit Size:
1 L
100mL
Pack of 3
This item is not returnable. View return policy

Product Code. 10496134

Brand: Thermo Scientific™ 37528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:
Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solution is ready to use; can be diluted further as needed
Safe—stable, thimerosal-free formulation

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PBS
Concentration 1X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line Blocker
Quantity 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.