Learn More
Thermo Scientific™ Blocker™ FL Fluorescent Blocking Buffer (10X)
Blocker FL Fluorescent Blocking Buffer is designed to block excess nonspecific binding sites to help reduce background fluorescence in western blotting applications.
Brand: Thermo Scientific™ 37565
87.33 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Blocker FL buffer helps enhance fluorescent protein detection by increasing the signal-to-noise ratio in blotting applications where background fluorescence may otherwise interfere with signal detection.
Compare and view all available blocking buffers
Working reagent is prepared by adding 1 part Blocker FL buffer to 9 parts ultrapure water. Optimal concentration should be determined empirically for each system. Compatible with both nitrocellulose and PVDF membranes.
Specifications
Blocker FL | |
Liquid | |
Blocker™ | |
Store in refrigerator (2°C to 8°C). |
100 mL | |
Western Blot | |
10X | |
Wet Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.