Learn More
Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium
Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote hemolysis.
£38.55
Product Code | Brand | Quantity | Format | Includes | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Includes | Manufacturing Origin | Price | Quantity & Availability | ||
10362223
|
Thermo Scientific™
PB5012A |
10/Pk. | 90mm Monoplate | TSA with Sheep Blood | Germany |
£38.55
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Grow a wide variety of fastidious organisms on Thermo Scientific™ Blood Agar (TSA with Sheep Blood), a general purpose non-selective medium with the addition of sheep blood in the formulation to promote the presence and visualization of haemolytic reactions so it can be used as a diagnostic tool.
Tryptone Soya Agar (casein soya bean digest agar) with defibrinated sheep blood is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown either in deep cultures or by incubation under anaerobic conditions. The combination of casein digest and papaic digest of soybean meal leads to optimal growth due to synergy between the protein supply of casein and the carbohydrate supply of soybeans. Sterile defibrinated sheep blood used to enrich the medium produces the hemolysis characteristics of different bacteria, such as streptococci, listeria, hemolytic Staphylococcus, Escherichia coli and Pseudomonas.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Blood Agar | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 8 Weeks