Learn More
BD Blood Collection Set
Intended for blood collection and short-term infusion
Brand: BD 367326
This item is not returnable.
View return policy
Description
Push button without luer adapter button ensures irreversible needle lock green 19mm, tubing length 305mm 21G gauge BD Vacutainer
Specifications
Intended for blood collection and short-term infusion | |
19.05 mm | |
Sterile |
Yes | |
Blood Collection Set |
Product Content Correction
Product Title
BD Blood Collection Set >
Spot an opportunity for improvement?Share a Content Correction