missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Discs

Product Code. 15922960 Shop All Sartorius Products
Click to view available options
Diameter (Metric) Outer:
22 mm
25 mm
150 mm
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 15922960

Brand: Sartorius Stedim Biotech™ FT3520022

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3 Discs are highly pure and ideal for blotting chromatography.

  • This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Made of high-purity cotton linters for uniform buffer flow and resulting blots.
  • No additives to avoid any interference during transfer.
  • BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.
TRUSTED_SUSTAINABILITY

Specifications

Description Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk
Diameter (Metric) Outer 22 mm
Film Thickness 0.76 mm
Format Disc
Grade BF 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.