missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Discs
Shop All Sartorius Products
Click to view available options
Diameter (Metric) Outer:
22 mm
25 mm
150 mm
Unit Size:
Pack of 100
Pack of 1000
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.
Specifications
Specifications
| Description | Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk |
| Diameter (Metric) Outer | 25 mm |
| Film Thickness | 0.76 mm |
| Format | Disc |
| Grade | BF 3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction