missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets are highly pure and ideal for blotting chromatography.
£83.10 - £128.00
Specifications
Description | Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk |
---|---|
Film Thickness | 0.76 mm |
Format | Sheet |
Grade | BF 3 |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15982800
|
Sartorius Stedim Biotech™
FT-2-520-135155N |
135 x 155 mm |
£83.10
Pack of 100 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
15992800
|
Sartorius Stedim Biotech™
FT-2-520-200200N |
200 x 200 mm |
£128.00
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.
Specifications
Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk | |
0.76 mm | |
Sheet | |
BF 3 |