missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets are highly pure and ideal for blotting chromatography.
£40.30 - £44.60
Specifications
Description | Capillary rise: 160mm/10 min., Weight: 550 g/m2, 100/Pk |
---|---|
Film Thickness | 1.30 mm |
Format | Sheet |
Grade | BF 4 |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15902810
|
Sartorius Stedim Biotech™
FT-2-521-110170G |
110 x 170 mm |
£40.30
Pack of 25 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15912810
|
Sartorius Stedim Biotech™
FT-2-521-150150G |
150 x 150 mm |
£44.60
Pack of 25 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 4 grade is ideal for transferring DNA or RNA according to the Southern technique or semidry blotting of proteins.
Specifications
Capillary rise: 160mm/10 min., Weight: 550 g/m2, 100/Pk | |
1.30 mm | |
Sheet | |
BF 4 |