missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Blue Carrier™ Protein

Product Code. 11814071 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11814071

Brand: Thermo Scientific™ 77130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easily prepare effective immunogens with the highly-soluble, mollusk-derived hemocyanin and complete hapten-carrier conjugation kit.

Thermo Scientific™ Blue Carrier Protein is a highly-soluble, mollusk-derived hemocyanin formulated in convenient package sizes and a complete kit for carbodiimide (EDC-mediated) hapten-carrier conjugation via amine and carboxyl groups.

Blue Carrier Protein is a purified preparation of Concholepas concholepas hemocyanin (CCH). The large protein exhibits most of the same immunogenic properties as the popular carrier protein, keyhole limpet hemocyanin (KLH). However, its signficantly better solubility provides greater flexibility in immunogen preparation protocols by allowing a broader range of buffer and pH conditions for coupling peptides, proteins and other haptens using classical crosslinkers such as glutaraldehyde, Sulfo-SMCC and EDC.

Highlights:

  • Convenient packages – choose lyophilized Blue Carrier Protein or complete EDC conjugation kit

  • High-yield conjugation – each molecule of CCH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers

  • Validated quality – the immunogenic carrier protein is specially purified to ensure uniform chemical structure and solubility for consistent performance

  • Superior solubility – increased solubility compared to KLH allows Blue Carrier Protein to be used in a broader range of conjugation protocols to accommodate unusual or poorly soluble haptens

  • Highly immunogenic – provides essentially the same level of immunogenicity and phylogenetic distance as keyhole limpet hemocyanin (KLH)

Properties of Blue Carrier Protein:

  • Alternative names: Concholepas concholepas hemocyanin, CCH
  • Molecular weight: Whole molecule: ∽8,000,000; Subunits: ∽400,000 and ∽350,000
  • Absorption: maxima 280nm, 350nm and 558nm
  • Extinction coefficient (Abs. 1mg/mL solution): 1.4 at 280nm; 0.35 at 346nm
  • A280/A350 Ratio: ∽4.0
  • Copper/protein ratio: ∽16 Cu2+/protein molecule
  • Isoelectric point: 6.0
  • Solubility: Soluble in aqueous buffers pH 7.0 to 8.0
  • Stability: Stable in aqueous buffers (∽pH 7.2 without Ca2+ or Mg2+)

Includes:

Kit contains lyophilized Blue Carrier Protein (in MES buffer), EDC crosslinker, buffers and desalting columns to easily prepare ready-to-use immunogens for injection

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C. Shipped on ice.
Quantity 100 mg
Product Line Imject
Product Type Imject Blue Carrier Protein

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.