missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Blue Insulated Lab Ice Pan

Use with ice, dry ice, dry ice alcohol slurries and liquid nitrogen. Bel Art™ Blue Insulated Lab Ice Pan has a temperature range of –196 to 90°C°C.

Brand:  Bel-Art™ Scienceware™ M18848-9191

Product Code. 12328707

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Temperature range: -196°C to 90°C
  • Non-sweating, superior insulation properties
  • Leakproof
  • Chemical resistant
  • Floating tube racks available (float in water, dry ice slurries and liquid nitrogen) For use with dry ice, dry ice alcohol slurries and liquid nitrogen.
Specifications

Specifications

For use with ice, dry ice, dry ice alcohol slurries and liquid nitrogen
Insulated Lab Ice Pan
9 L
Foam Polymer
Blue
-196°C to +90°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers