missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Blue LED Transilluminator
GREENER_CHOICE

Blue LED Transilluminator equipped with a viewing area.

Brand:  Syngene™ BT-ULTRA-BRIGHT-LED

Product Code. 16544331

  • £1075.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Even blue epi LED illumination
  • High sensitivity with improved S/N ratio
  • Image a range of fluorophores- EtBr, SYBRSafe, GelGreen, SYBR Gold, SYBRGreen, SYPRORuby, SYPROOrange, GelRed
  • LED is safe to skin and eyes with no damage to DNA
Specifications

Specifications

Fixed
470 nm
Viewing area (20 x 16 cm)
LED Blue
LED Transilluminator
100/115/220/230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers