missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Color-coded Octahedral PTFE Stir Bar
Use when quick visual identification is highly important. Fisherbrand™ Color-coded Octahedral PTFE Stir Bar is available in three colors: blue, red, and yellow.
£2.97 - £49.40
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Description | PTFE encapsulated Alnico V magnet core |
Shape | Octahedral |
For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Color | Price | Quantity & Availability | |||
12574987
|
Fisherbrand™
|
8 mm | 15 mm | Blue |
£15.00
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11848912
|
Fisherbrand™
|
8 mm | 22 mm | Blue |
£28.60
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11522313
|
Fisherbrand™
|
8 mm | 38 mm | Blue |
£24.97
Pack of 10 |
In Stock
Log in to see stock. |
|||
11532313
|
Fisherbrand™
|
8 mm | 51 mm | Blue |
£32.80
Pack of 10 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||
11888912
|
Fisherbrand™
|
13 mm | 75 mm | Blue |
£28.10
Pack of 5 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||
11792145
|
Fisherbrand™
|
8 mm | 13 mm | Blue |
£12.20
Pack of 10 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
11512313
|
Fisherbrand™
|
8 mm | 25 mm | Blue |
£15.90
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11888902
|
Fisherbrand™
|
8 mm | 13 mm | Red |
£11.00
Pack of 10 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
11898902
|
Fisherbrand™
|
8 mm | 15 mm | Red |
£17.96
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11808912
|
Fisherbrand™
|
8 mm | 22 mm | Red |
£20.90
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11582303
|
Fisherbrand™
|
8 mm | 38 mm | Red |
£28.60
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11572303
|
Fisherbrand™
|
8 mm | 25 mm | Red |
£19.04
Pack of 10 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
|||
11963574
|
Fisherbrand™
|
13 mm | 75 mm | Red |
£32.10
Pack of 5 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11542313
|
Fisherbrand™
|
8 mm | 13 mm | Yellow |
£12.59
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11898912
|
Fisherbrand™
|
8 mm | 15 mm | Yellow |
£17.13
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11808922
|
Fisherbrand™
|
8 mm | 22 mm | Yellow |
£36.33
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11572313
|
Fisherbrand™
|
8 mm | 38 mm | Yellow |
£28.60
Pack of 10 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
11562313
|
Fisherbrand™
|
8 mm | 25 mm | Yellow |
£17.60
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
13573120
|
Fisherbrand™
|
13 mm | 75 mm | Yellow |
£36.07
Pack of 5 |
In Stock
Log in to see stock. |
|||
11582313
|
Fisherbrand™
|
8 mm | 51 mm | Yellow |
£49.40
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11828922
|
Fisherbrand™
|
8 mm | 38 mm | Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11818912
|
Fisherbrand™
|
8 mm | 25 mm | Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11828912
|
Fisherbrand™
|
8 mm | 38 mm | Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11868912
|
Fisherbrand™
|
8 mm | 38 mm | Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Color-coded, octagonal stirrer bars
- For use where identification is of prime importance
- Note: Colored PTFE coatings are not as inert as pure PTFE
Specifications
Autoclavable | |
PTFE encapsulated Alnico V magnet core | |
Containers & Vessels / Stirrer Hot Plates |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
Octahedral |