missing translation for 'onlineSavingsMsg'
Learn More

3M™ Blue Disposable Visitor Lab Coats, Series 4400

Use for hygiene applications. The 3M Disposable Visitor Lab Coats series is made from lightweight polypropylene.

Brand:  3M™ 7000089702

 View more versions of this product

Product Code. 12911335

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ideal for hygiene applications
  • 100% soft, breathable, and lightweight polypropylene
  • Zip fastener
  • Internal pocket
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lab Coat
Polypropylene
Zipper
Single-use
38 to 40 in.
CE Category 1
Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
X-Large
Blue
Elastic
Serged
96 to 101 cm
Non-hazardous liquids and dust
Individual packaging
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers