missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color-coded Micro PTFE Stir Bar

Use when quick visual identification is highly important. Fisherbrand™ Color-coded Micro PTFE Stir Bar is available in three colors: blue, red, and yellow.

Brand:  Fisherbrand™ 001.815-Y

 View more versions of this product

Product Code. 11567193

  • £13.40 / Pack of 10
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Color-coded, micro stirrer bars
  • For use where identification is of prime importance
  • Note: Colored PTFE coatings are not as inert as pure PTFE
Specifications

Specifications

Autoclavable
Yellow
Containers & Vessels / Stirrer Hot Plates
1.5 mm
5/Pk.
Stir Bar
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
PTFE
15 mm
Micro
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers