Learn More
Syngene™ Blue Slimline LED Transilluminator with Sliding Tray
GREENER_CHOICE
LED Transilluminator suitable for a variety of imaging needs.
Brand:  Syngene™ GX-ULTRA-SLIM-LED
Product Code. 16574331
  • £725.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Even blue epi LED illumination
  • High sensitivity with improved S/N ratio
  • Image a range of fluorophores- EtBr, SYBRSafe, GelGreen, SYBR Gold, SYBRGreen, SYPRORuby, SYPROOrange, GelRed
  • LED is safe to skin and eyes with no damage to DNA
Specifications
Specifications
470 nm
Transilluminator
Sliding tray
7 x 12 cm