Learn More
BMP1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547222
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) TLL-2 is observed, and less than 1% cross-reactivity with rhBMP-2, -3, -3b, -4, -5, -6, -7, -8, -9, -10, and -15 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
BMP1 Polyclonal antibody specifically detects BMP1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
BMP1 | |
Polyclonal | |
Unconjugated | |
BMP1 | |
BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD | |
Goat | |
Antigen affinity chromatography | |
RUO | |
649 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P13497 | |
BMP1 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-1/PCP Ala121-Gln730 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |