missing translation for 'onlineSavingsMsg'
Learn More

BMS 193885, Tocris Bioscience™

Product Code. 18054652
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, competitive NPY Y1 receptor antagonist

Potent, competitive neuropeptide (NPY) Y1 antagonist (Ki = 3.3 nM, IC50 = 5.9 nM) that displays > 47, > 100, > 160, > 160 and > 160-fold selectivity over σ1, α1, Y2, Y4 and Y5 receptors respectively. Reduces food intake and body weight via central Y1 inhibition and is brain penetrant.

Chemical Identifiers

CAS 679839-66-8
Molecular Formula C36H48N4O9
Molecular Weight (g/mol) 680.80
MDL Number MFCD16878998
InChI Key BYHJIPSVXAFCDI-UHFFFAOYNA-N
Synonym 1,4-dihydro-4-3-3-4-3-methoxyphenyl-1-piperidinyl propyl amino carbonyl amino phenyl-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester l-lactate
PubChem CID 56972234
IUPAC Name 2-hydroxypropanoic acid; 3,5-dimethyl 4-{3-[({3-[4-(3-methoxyphenyl)piperidin-1-yl]propyl}carbamoyl)amino]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC(O)C(O)=O.COC(=O)C1=C(C)NC(C)=C(C1C1=CC(NC(=O)NCCCN2CCC(CC2)C2=CC(OC)=CC=C2)=CC=C1)C(=O)OC

Specifications

Quantity 10 mg
Formula Weight 680.79
Percent Purity >99%
Chemical Name or Material BMS 193885
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'