missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 193885, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, competitive neuropeptide (NPY) Y1 antagonist (Ki = 3.3 nM, IC50 = 5.9 nM) that displays > 47, > 100, > 160, > 160 and > 160-fold selectivity over σ1, α1, Y2, Y4 and Y5 receptors respectively. Reduces food intake and body weight via central Y1 inhibition and is brain penetrant.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 680.79 |
Percent Purity | >99% |
Chemical Name or Material | BMS 193885 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'